missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Rectangular Plate, L x W: 100 x 50mm, Thickness: 4mm, Thermo Scientific™

Catalog No. AA33158KF
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33158KF 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA33158KF Supplier Thermo Scientific Chemicals Supplier No. 33158KF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Al-23 Rectangular Plate
Quantity 1 Ea.
Form Plate