missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Fish Spine Beads, O.D.: 5mm, Bore: 2.5mm, Length: 5mm, Thermo Scientific™

Catalog No. AA32907KX
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA32907KX 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA32907KX Supplier Thermo Scientific Chemicals Supplier No. 32907KX

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Name or Material Al-23 Fish Spine Beads
Quantity 100 Pk.
Form Fish Spine Beads