Learn More
Al-23 Combustion Tray, L1: 50mm, L2: 48mm, H: 20mm, W: 3mm, Vol.: 33, Thermo Scientific™
Catalog No. AA33184KF
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
AA33184KF 1 Ea.
1 options
Catalog No. AA33184KF Supplier Thermo Scientific Chemicals Supplier No. 33184KF
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Chemical Name or Material Al-23 Combustion Tray
Quantity 1 Ea.