missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Boat;L1 (mm), 130;L2 (mm), 116;B1 (mm), 31;B2 (mm), 27;H (mm), 17;W (mm), 3;Vol, 36.0

Catalog No. AA33182KF
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33182KF 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA33182KF Supplier Thermo Scientific Chemicals Supplier No. 033182KF

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Al-23 Boat
Quantity 1 Pc.
Form Boat

RUO – Research Use Only