Learn More
Enzo Akt monoclonal antibody (5C10) (100g)
Catalog No. 502009943
Encompass
Clone: 5C10. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to C-terminal human Akt (protein kinase B; PKB) conjugated to KLH. Species reactivity: Human, Mouse, Rat, Dog. Specificity: Recognizes a C-terminal epitope (aa 466-480) of phosphorylated and non-phosphorylated Akt isoforms (see Technical Note below). Applications: ELISA, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 502009943 Supplier Enzo Supplier No. ALX804553C100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clone: 5C10. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to C-terminal human Akt (protein kinase B; PKB) conjugated to KLH. Species reactivity: Human, Mouse, Rat, Dog. Specificity: Recognizes a C-terminal epitope (aa 466-480) of phosphorylated and non-phosphorylated Akt isoforms (see Technical Note below). Applications: ELISA, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.