missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Akro-Mils™ AkroBins™ Medium Storage Bin Accessories: Bin Lids
Description
- See-through crystal styrene
- Assembly required; hinge pins included
Warranty
One-year limited
Specifications
Specifications
| Type | Bin Lid |
| Color | Clear |
| Includes | Hinge Pin |
| For Use With (Equipment) | 30235 Bin |