missing translation for 'onlineSavingsMsg'
Learn More

Akro-Mils™ Akro-Grid Storage Totes: Lids

Catalog No. 08757604
Encompass
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
08757604 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08757604 Supplier Akro-Mils™ Supplier No. 33061

For use with Akro-Grid Storage Totes 08-757-605, 08-757-601 and 08-757-651

Specifications

Type Bin Lid
Material Plastic
Color Clear
For Use With (Equipment) 33164 Bin, 33166 Bin, 33168 Bin