missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ AKAP1 Recombinant Protein Antigen
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human AKAP1. The AKAP1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 8165 |
| Species | Human |
| Product Line | Novus |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | AKAP1 |
| Molecular Weight (g/mol) | 33 kDa |
| Quantity | 100 μL |
| Show More |