missing translation for 'onlineSavingsMsg'
Learn More

AK Protective Benchtop Shields

Catalog No. 17987140D
Change view
Click to view available options
Width (Metric):
30.4 cm
40.6 cm
Thickness (Metric):
0.63 cm
0.95 cm
Width (English):
12 in.
16 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric) Thickness (Metric) Width (English)
17987140D 30.4 cm 0.63 cm 12 in.
17987140B 30.4 cm 0.95 cm 12 in.
17987140C 40.6 cm 0.63 cm 16 in.
17987140A 40.6 cm 0.95 cm 16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17987140D Supplier AK Supplier No. AK 204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These durable acrylic shields are ideal for use with blood, liquids, culture specimens, or other hazardous materials. The angled top protects the face against splash and flying particles, while the patented curved-side design allows more natural arm movement, so workers can more easily reach the work surface.Shields are available in two thicknesses: 3/8 and 1/4 in. (0.95 and 0.6cm). The 3/8 in. thick (0.95cm) shield offers protection against radiation exposure when used with low-level beta-emitting isotopes such as 32P. The 1/4 in. thick (0.6cm) shield should be used for splash protection only. Both shields offer a choice of two work surface sizes: 12L × 12 in.W (31 × 31cm) or 12L × 16 in.W (31 × 41cm). All shields are 20 in. high (51cm).

Specifications

Thickness (Metric) 0.63 cm
Width (English) 12 in.
Width (Metric) 30.4 cm
Height (English) 20 in.
Height (Metric) 50.8 cm
For Use With (Equipment) For use with blood, liquids, culture specimens, or other hazardous materials