Learn More
AK Single Compartment Finger Cot Dispenser
Catalog No. 17987140X
Change view
Click to view available options
Single
Catalog No.
17987140X Single
1 options
Catalog No. 17987140X Supplier AK Supplier No. AK 901C
Only null left
Ideal for dispensing and organizing finger cots in production facilities. Unit features a hinged lid, and bold writing for easy product identification.
  • Constructed of clear, impact-resistant 1/8 in. PETG
  • Unit features a hinged lid and doors for easy loading and removal
  • Dispenser can be wall-mounted or free-standing
  • Bold writing for easy product identification
Specifications
Product Type Finger Cot Dispenser
Color Clear
Dimensions (D x W x H) Interior 8.5 x 6 x 11.25 in.
Holds Finger Cots
Material PETG
No. of Compartments 1
Product Style Top Loading
Thickness (English) 0.125 in.