missing translation for 'onlineSavingsMsg'
Learn More

AK Acrylic Ambi-Glove Dispenser

Catalog No. 17987140V
Change view
Click to view available options
Color:
Amber
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17987140V Amber
17987140W Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17987140V Supplier AK Supplier No. AK 7941A

Specially designed for ambidextrous gloves, with a front access hole for convenient product dispensing.

  • Designed for storing and dispensing ambidextrous gloves
  • Front opening for convenient product access
  • Amber color helps prevent ozone degradation of latex gloves

Specifications

Product Type Glove Dispenser
Color Amber
Dimensions (D x W x H) Interior 15 x 11.5 x 7.75 in.
Holds Gloves
Material Acrylic
No. of Compartments 1
Product Style Top Loading
Thickness (English) 0.25 in.