Learn More
AirClean™ Systems AirMax™ Polypropylene Total Exhaust Fume Hoods
Description
- Manufactured from all-white, flame-retardant polypropylene
- NO RUST- all contact surfaces are constructed of polypropylene
- Microprocessor controller monitors face velocity and displays the real-time airflow on the LCD screen in linear feet per minute
- Vertical sliding safety glass sash
- Built-in vapor-proof fluorescent light
- Easy-access utilities panel for operation and maintenance
- Non-hydroscopic, will not absorb moisture
- Large volume spill basin with solid work surface
Safety Features:
- Advanced airfoil design that is important to the overall safety and performance of the fume hood
- Rolled entry and advanced air foil design prevents reverse flow and eddy currents from escaping the fume hood
- Double sidewall construction allows mounting of services and outlets on the posts
- Effective spill containment- Removable work surface exposes spill basin that may be fitted with drain for disposal of any spills
- Removable work surface exposes spill basin that may be fitted with drain for disposal of any spills
- Rear baffle extends below work surface to effectively remove any heavy gases or evaporation from a spill
- Can be fitted with wet scrubber system to remove high volumes of water soluble acids and solvents
- Auto dosing system for neutralization of acids
- Polycarbonate sash for HF applications
- Polypropylene blowers
- Variable speed driver to make a true VAV system
- Polypropylene base cabinets
Specifications
Specifications
| Product Type | Total Exhaust Fume Hood |
| Dimensions (D x W x H) Interior | 21 x 85 x 32 in. |
| Liner Material | Polypropylene |