Learn More
AirClean™ Systems PharmaSafe™ Ductless Class II Enclosure Accessory, Cart
Description
- For use with PharmaSafe™ Ductless Class II Enclosure
- Material: Steel
Specifications
Specifications
| Product Type | Cart |
| For Use With (Equipment) | ACPHS36 and ACPCR36 model units with width: 91.4cm (36 in.) |
| Height (Metric) | 86.4 cm |
| Material | Steel |