Learn More
AirClean™ Systems AC600 Series Vertical Laminar Flow Fume Hood
For applications that are not hazardous to operator but require clean work area, choose an Airclean AC600 Series Vertical Laminar Flow Fume Hood.
Supplier:  AirClean™ Systems AC648LFC
Catalog No. 360993861
Only null left
Description
Description
  • UVTect™ microprocessor controller
  • Class 100 (ISO 5) vertical laminar flow
  • 360-degree visibility
  • Integral polypropylene base for easy cleaning
  • Supplied fully assembled
  • Built-in fluorescent light
  • HEPA filter monitor automatically indicates when filter change is required
  • Available in 110V or 220V versions
  • Compact, light weight design allows for multiple “clean zones” within laboratory
Options
  • Shortwave UV light
  • Custom sturdy cart allows unit to be made portable
  • Polypropylene - metal free construction
  • ULPA filter
  • Sliphatch access port
Certifications
UL listed, CSA, CE marked
Compliance
Certified to meet or exceed applicable clean bench standards.
Product Suggestions
Product Suggestions