Learn More
AirClean™ Systems Polypropylene Ductless Fume Hood, Sliding Sash
Help keep your employees well-protected. AirSafe™ Polypropylene Ductless Fume Hood features an automatic safety controller and vertical sliding safety glass sash.
Supplier:  AirClean™ Systems AC4000S
 View more versions of this product
Catalog No. 361018797
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alert:
Requires an air filter in order to operate. Air filter must be purchased separately.
Description
Description
  • AirSafe™ automatic safety controller monitors airflow face velocity and carbon filter saturation
  • Vertical sliding safety glass sash
  • AirZone™ internal baffling promotes even airflow for maximum containment
  • Access panels on both side of hood allows field service of all fixtures
  • FlowSmooth™ airfoil on sash lip promotes laminar airflow
  • Dual wall construction allows for front mounting of services
  • Chemical resistant polypropylene construction
  • Activated bonded carbon, impregnated chemisorptive and HEPA filters for containment of virtually any toxic vapor, fume, gas, or particulate
  • Integral vapor-proof fluorescent lighting system
  • Available in 110V or 220V AC versions
  • No installation required, shipped fully assembled
Options
  • Sturdy mobile cart
  • Vented and unvented polypropylene base cabinets
  • Remote services - gas, water, vacuum, and air
  • Duplex electrical outlets
  • Polypropylene sinks and water fixtures
  • Slip hatch electrical access port
Specifications
Chemical Identifiers
Polypropylene
60 in.
110/220 V
OSHA, ANSI Z9.5, ASHRAE 110
152.4 cm
Specifications
Polypropylene
63.5 x 111.7 x 91.4 cm
152.4 cm
50 in.
OSHA, ANSI Z9.5, ASHRAE 110
60 in.
110/220 V
Product Suggestions
Product Suggestions