missing translation for 'onlineSavingsMsg'
Learn More

Air Systems™ Venturi-Style Pneumatic Air Blowers

Catalog No. 18502
Encompass
Change view
Click to view available options
Diameter (English) Fan:
7.37 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Fan
18502 7.37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18502 Supplier Air Systems™ Supplier No. ASI 1200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aluminum base and galvanized steel horn. Expel explosive and hazardous vapors or fumes from underground spaces. No moving parts—horns operate on compressed air or steam. Horn 18-503 meets ventilation requirements for underground storage tanks. Static grounding lug is standard on all models. Horns stand on lightweight aluminum base.

Specifications

Diameter (English) Fan 7.37 in.
Product Type Venturi pneumatic air blower
Flow Rate 38 cfm at 50 psig
Length (English) 32.12 in.
Length (Metric) 81.58 cm
Weight (English) 8.1 lb.
Weight (Metric) 4 kg