missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Air High Performance Pipe Marker, 0.375 to 0.5 in., White on Blue
Description
- Designed for use on rusty, dirty, wet or rough pipes where pressure sensitive labels aren’t effective.
- Resistant to UV light exposure, high humidity, high temperatures, abrasion, chemical reagents and physical agitation.
- Self-adhesive end strip adheres to the marker's body to ensure a tight, continuous wrap.
- Excellent chemical resistance.
Specifications
Specifications
| Adhesive Type | Non-adhesive |
| Coating | Laminate |
| Color | White on Blue |
| Display | Text |
| Finish | Gloss |
| Format | Sheet |
| Material | Coiled Plastic (B-689) |
| Product Type | Pipe Marker |
| Sterility | Non-sterile |
| Dimensions (L x W) | 4 x 4 in. |
| Show More |