Learn More
Air Core Solenoid
Catalog No. S94209
Change view
Click to view available options
Material:
Copper
Catalog No. Material
S94209 Copper
1 options
Catalog No. S94209 Supplier Customer Channels Group (Ccg) Supplier No. EPD10141
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provides a useful magnetic field for many magnetism and electricity experiments.
  • Investigate the mass of electron apparatus and experiments using the current balance (S94280)
  • Constructed using four layers, approximately 710 turns, of #23 gauge (0.67mm) polyurethane-enameled copper wire
  • Coil is designed to carry 7-10A intermittently and will sustain a current of 5A
To use:
  • Connect a power supply that does not put out more than 5A to the solenoid
  • Coil will generate a magnetic field when current passes through it
  • Use a compass, iron rod, or iron filings to study the nature of the generated electric field
Specifications
Material Copper
For Use With (Application) Provides useful magnetic field for magnetism, electricity experiments
Product Type Air Core Solenoid