missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ AIN-76A Rodent Diet

Catalog No. p-4185819
Encompass
Change view
Click to view available options
Particle Shape:
Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Shape
14726615 Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14726615 Supplier Bio-Serv™ Supplier No. F0761
Only null left

Chlorophyll free—does not interfere with imaging studies

  • Purified and extensively used in research
  • Balanced to meet nutritional needs of rodents
  • Phytoestrogen free
  • Nutritionally assayed
  • Ingredients: Sucrose, Dextrose, Casein, Corn Starch, Corn Oil, Cellulose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Bitartrate

Specifications

Product Type Rodent Diet
Components Sucrose, Casein, Corn Starch, Corn Oil, Cellulose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Bitartrate
Format Powder
For Use With (Application) Used in research
Particle Shape Powder
Quantity 5000 g
Species Rodent