missing translation for 'onlineSavingsMsg'
Learn More

Germaine™ Laboratories AimTab™ Reducing Substances Tablets

Catalog No. 23111358
Change view
Click to view available options
Quantity:
36 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23111358 36 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23111358 Supplier Germaine™ Laboratories Supplier No. 13336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the semi-quantitative determination of the amount of reducing substances (glucose, lactose, fructose, galactose, pentoses) in urine, stool, wine, or any other liquids. Tablets provides clinically useful information on carbohydrate metabolism. FOR FORENSIC USE ONLY.

  • Color chart included for easy to read results
  • Can be used to perfom the 5-Drop Method or 2-Drop Method
  • Store Between 15° to 30°C
  • Do not store in sunlight or humid places

Specifications

Content And Storage 15°C to 30°C
CE Marker No
Detectable Analytes Glucose, Lactose, Fructose, Galactose, Pentose
For Use With (Application) For urine, stool or other liquids.
Packaging Type Bottle
Quantity 36 Tablets