missing translation for 'onlineSavingsMsg'
Learn More

Germaine™ AimStrip™ Xylazine Urine/Powder Dipstick, 25/Kit

Catalog No. 23111435
Change view
Click to view available options
Detectable Analytes:
Xylazine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23111435 Xylazine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23111435 Supplier Germaine™ Supplier No. 43347

The AimStrip™ Xylazine Urine/Powder Dipstick is rapid test for the qualitative detection of xylazine in human urine and in powders.

  • A rapid test for the qualitative detection of xylazine in human urine and powder.
  • For forensic use only, not for in vitro diagnostic use.
  • Rapid chromatographic immunoassay for the detection of xylazine in human urine and powder.
  • Cut-off concentration of 1,000 ng/mL.
  • Results in 5 minutes.
  • No instrument required.
  • The test utilizes the antibody to selectively detect elevated levels of xylazine in urine and powder.
  • XYLAZINE has also been reported to be misused as a horse doping agent, a drug of abuse, and as source of accidental or intended poisonings.

For powder samples, only a small 1 to 10 mg sample size is needed to be diluted with water and perform the test. Confirm powder sample results at 1 minute, and urine sample results at 5 minutes.

Specifications

CE Marker No
Clia Complexity Not Applicable
Detectable Analytes Xylazine
Format Test Strips
Sample Type Urine, Powder
Sensitivity Cut off concentration of 1000 ng/mL
Type Test Strips
For Use With (Application) Drug Panel (Urine)
Includes 25 Test Strips/Kit, and a package insert.
Test Time Confirm powder sample results at 1 minute, and urine sample results at 5 minutes.

For Forensic Use Only