Learn More
AIMS™ Stereotaxic Magnifier
Catalog No. 14370251
Encompass
Change view
Click to view available options
Product Type:
Magnifier, Stereotaxic
On-Off Switch for Stereotaxic Magnifier
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14370251 On-Off Switch for Stereotaxic Magnifier
11900101 Magnifier, Stereotaxic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14370251 Supplier AIMS™ Supplier No. LLSW
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
With flex arm and non-obstructive rimless magnifying glass
  • Uses magnetic base/puck system to attach to non-metallic surfaces
  • Can also be directly attached to metallic surfaces
  • Allows continuous eye contact when moving into magnification area
  • Magnification: 2.3X (main lens), 4X (insert lens)
Specifications
For Use With (Application) Magnification activities
Product Type On-Off Switch for Stereotaxic Magnifier