Learn More
AIMS™ Humane Mouse Restrainer
Accessory for AIMS tattoo products.
Supplier:  AIMS™ 1000
Encompass
Catalog No. 14370147
Only null left
Description
Description
  • Requires less pressure and makes it easier to hold the mouse tail for tattooing
  • Features three holes, small, medium and large, to accommodate different tail sizes
Specifications
Specifications
Humane Mouse Restrainer
Product Suggestions
Product Suggestions