missing translation for 'onlineSavingsMsg'
Learn More

AIMS™ Body-Contour Cone-Shaped Plastic Disposable Rodent Restraints

Catalog No. 14370110
Encompass
Change view
Click to view available options
Color:
Clear
Rose
Size:
Small
Weight Range:
≤120 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Weight Range
14370110 Clear Small ≤120 g
14370113 Rose Small ≤120 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14370110 Supplier AIMS™ Supplier No. 1801CV

Includes

Instructions and twist ties

Humane, safe, disposable rodent restraint system

Available in a variety of sizes and colors. Cone-shaped bags provide easier animal capture and increased animal comfort.

  • Precut sizes for quick use
  • Easy-Flex-PETM provides effortless wrapping and added animal comfort
  • Body-Contour cone shape provides easier restraint, exceptional comfort, and reduced animal stress
  • Black and rose color bags reduce natural light and aids in reducing animal agitation and stress

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.

Specifications

Color Clear
For Use With (Application) Tail Tattooing, Tail Vein Injections, Oral Dosing, Decapitation and General Restraint
Type Rodent Restraint
Disposable Single-use
Includes Instructions, twist ties
Material Plastic
Shape Cone
Size Small
Weight Range ≤120 g