Learn More
AIMS™ ATS-3 General Rodent Tattoo System Needles
Developed to minimize animal pain and tissue damage during tattooing
Supplier: AIMS™ 3035
Description
Accessory for ATS-3 General Rodent Tattoo System
- Made of 401 stainless steel and silver solder
- Pre-sterilized and ready for tattooing
- Proper needles allows for humane and rapid animal tattooing
- Tails are not damaged
- 5 needles per pack
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Chemical Identifiers
| Tattooing | |
| ATS-3 general rodent tattoo system | |
| Sterile |
| Tattoo Needle | |
| 401 Stainless Steel, Silver Solder |
Specifications
| Tattooing | |
| ATS-3 general rodent tattoo system | |
| Sterile |
| Tattoo Needle | |
| 401 Stainless Steel, Silver Solder | |
| Ultra Fine Point |