Learn More
AIMS™ ATS-3 General Rodent Tattoo System Needles
Catalog No. 14370152
Encompass
Change view
Click to view available options
Tip Style:
Heavy Duty Point
Standard Point
Ultra Fine Point
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
14370152 Standard Point
14370153 Ultra Fine Point
14370154 Heavy Duty Point
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14370152 Supplier AIMS™ Supplier No. 3005
Developed to minimize animal pain and tissue damage during tattooing
Accessory for ATS-3 General Rodent Tattoo System
  • Made of 401 stainless steel and silver solder
  • Pre-sterilized and ready for tattooing
  • Proper needles allows for humane and rapid animal tattooing
  • Tails are not damaged
  • 5 needles per pack
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Specifications
For Use With (Application) Tattooing
Type Tattoo Needle
For Use With (Equipment) ATS-3 general rodent tattoo system
Material 401 Stainless Steel, Silver Solder
Sterility Sterile
Tip Style Standard Point