missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AIM V™ Medium (research grade), AlbuMAX™ Supplement

Catalog No. 31035025
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31035025 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31035025 Supplier Gibco™ Supplier No. 31035025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AIM V™ Medium (research grade), AlbuMAX™ Supplement

Contains AlbuMAX™ supplement (bovine serum albumin) substituted for human serum albumin. Contains L-glutamine, streptomycin sulfate at 50μg/mL, and gentamicin sulfate at 10μg/mL.

Specifications

Cell Type Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
Form Liquid
Product Type AIM V™ Medium
Sterility Sterile-filtered
With Additives Glutamine, Phenol Red, AlbuMAX
Product Line AIM V
Quantity 500 mL
Shipping Condition Room Temperature
Included Antibiotics Streptomycin Sulfate, Gentamicin
Purity or Quality Grade Research Grade
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less
Is any supplementation needed for AIM V Medium (research grade), AlbuMAX Supplement (Cat. No. 31035025)?

There is no extra supplementation required for this product, but we do recommend refreshing L-glutamine supplementation every 30 days.

For Research Use Only. Not for use in diagnostic procedures.