missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AIM V™ Medium, liquid (research grade)

Catalog No. 12055091
Encompass
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12055091 500 mL
12055083 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12055091 Supplier Gibco™ Supplier No. 12055091

missing translation for 'validMainframeMsg'

AIM V (research grade) is a medium designed for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells.

AIM V (research grade) is a medium designed for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells.

AIM V Medium contains L-glutamine, streptomycin sulfate at 50 μg/mL, and gentamicin sulfate at 10 μg/mL.

Specifications

Cell Type Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
Form Liquid
Product Type Serum Free Medium (SFM)
Sterility Sterile-filtered
With Additives Glutamine, Phenol Red, HSA
Product Line AIM V
Quantity 500 mL
Shipping Condition Room Temperature
Included Antibiotics Streptomycin Sulfate, Gentamicin
Purity or Quality Grade Research Grade
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less
What is the difference between AIM V Medium (Cat. Nos. 12055083, 12055091) and CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK)?

The difference is in the intended usage of the two media. AIM V Medium (Cat. Nos. 12055083, 12055091) is for research use only whereas CTS AIM V SFM (Cat. Nos. 0870112DK, 0870112BK) is for research use or manufacturing of cell, gene, or tissue-based products.

Which serum-free medium would you recommend for culture and expansion of mouse T cells?

You can try using AIM V Medium (Cat. No. 12055083 or 12055091) for growth of mouse T cells. For mouse cells, we recommend adding 50 µM beta-mercaptoethanol (Cat. No. 21985023).

For Research Use Only. Not for use in diagnostic procedures.