missing translation for 'onlineSavingsMsg'
Learn More

Agomelatine 98.0+%, TCI America™

Catalog No. A26061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26061G Supplier TCI America Supplier No. A26061G

Chemical Identifiers

CAS 138112-76-2
Molecular Formula C15H17NO2
Molecular Weight (g/mol) 243.306
MDL Number MFCD00916659
InChI Key YJYPHIXNFHFHND-UHFFFAOYSA-N
Synonym N-[2-(7-Methoxy-1-naphthalenyl)ethyl]acetamide
PubChem CID 82148
IUPAC Name N-[2-(7-methoxynaphthalen-1-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=CC=CC2=C1C=C(C=C2)OC

Specifications

Melting Point 110°C
Color White
Quantity 1 g
Formula Weight 243.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Agomelatine
RTECSNumber AC5956323
TSCA No
Recommended Storage Refrigerator