missing translation for 'onlineSavingsMsg'
Learn More

Agmatine Sulfate 98.0+%, TCI America™

Catalog No. A0310100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0310100MG 100 mg
A03101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0310100MG Supplier TCI America Supplier No. A0310100MG

Chemical Identifiers

CAS 2482-00-0
Molecular Formula C5H16N4O4S
Molecular Weight (g/mol) 228.267
MDL Number MFCD00013109
InChI Key PTAYFGHRDOMJGC-UHFFFAOYSA-N
Synonym agmatine sulfate, 1-4-aminobutyl guanidine sulfate, 1-amino-4-guanidinobutane sulfate salt, 4-aminobutyl guanidinium sulphate, unii-ru0176ql8i, n-4-aminobutyl guanidine sulfate salt, agmatine; sulfuric acid, 4-guanidinobutylamine sulfate salt, guanidine, 4-aminobutyl-, sulfate 1:1
PubChem CID 2794990
IUPAC Name 2-(4-aminobutyl)guanidine;sulfuric acid
SMILES C(CCN=C(N)N)CN.OS(=O)(=O)O

Specifications

Melting Point 231°C
Color White-Yellow
Quantity 100 mg
Formula Weight 228.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Agmatine Sulfate
EINECSNumber (2)-3985
TSCA No