missing translation for 'onlineSavingsMsg'
Learn More

Agmatine sulfate, 97%

Catalog No. p-7036903
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55363MD 250 mg
AAH5536303 1 g
AAH5536306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55363MD Supplier Thermo Scientific Chemicals Supplier No. H55363MD

CAS: 2482-00-0 | C5H16N4O4S | 228.267 g/mol

Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

Solubility
Soluble in water, nearly insoluble in alcohol

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 2482-00-0
Molecular Formula C5H16N4O4S
Molecular Weight (g/mol) 228.267
MDL Number MFCD00013109
InChI Key PTAYFGHRDOMJGC-UHFFFAOYSA-N
Synonym agmatine sulfate, 1-4-aminobutyl guanidine sulfate, 1-amino-4-guanidinobutane sulfate salt, 4-aminobutyl guanidinium sulphate, unii-ru0176ql8i, n-4-aminobutyl guanidine sulfate salt, agmatine; sulfuric acid, 4-guanidinobutylamine sulfate salt, guanidine, 4-aminobutyl-, sulfate 1:1
PubChem CID 2794990
IUPAC Name 2-(4-aminobutyl)guanidine;sulfuric acid
SMILES C(CCN=C(N)N)CN.OS(=O)(=O)O

Specifications

Melting Point 231°C to 238°C
Linear Formula H2N(CH2)4NHC(=NH)NH2·H2SO4
Quantity 250 mg
Beilstein 3918807
Merck Index 14,188
Solubility Information Soluble in water, nearly insoluble in alcohol
Formula Weight 228.27
Percent Purity 97%
Chemical Name or Material Agmatine sulfate
EINECSNumber 219-617-3
RTECSNumber ME8413000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only