Learn More
Brady™ Aggressive Adhesive Multipurpose Polyester Labels
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady™ handheld label printer.
Supplier:  Brady™ M490422
 View more versions of this product
Catalog No. 17012199
Description
Description
  • Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Specifications
Chemical Identifiers
UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863
White
Polyester
Asset tracking;Barcode labelling;Component and equipment labelling;Data and telecommunications labelling;General identification;Inventory and inspection labelling
-40°C to 100°C
0.102 mm
Permanent Acrylic
Gloss
Non-sterile
-40°F to 212°F
0.004 in
Specifications
UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863
White
Die-cut
Label
Asset tracking;Barcode labelling;Component and equipment labelling;Data and telecommunications labelling;General identification;Inventory and inspection labelling
180
-40°C to 100°C
0.102 mm
0.75 in.
19.05 mm
38.10 mm
Permanent Acrylic
Gloss
Polyester
Non-sterile
BMP41;BMP51;BMP53;M511;M410;M510
-40°F to 212°F
0.004 in
19.05 mm (W) x 38.10 mm (H)
0.75 in
1.5 in
M4-90-422
Product Suggestions
Product Suggestions