Learn More
Beckman Coulter Canada Lp RNAClean XP, 40 mL
Supplier: Beckman Coulter A63987
RNAClean XP 40 mL reagent kit provides a simple, flexible and highly reproducible method for purifying nucleic acid products generated in common enzymatic reactions such as cDNA synthesis and in vitro transcription (IVT) reactions.