missing translation for 'onlineSavingsMsg'
Learn More

Electron Microscopy Sciences Agarose Powder 100 GR

Catalog No. 50980167
Encompass

CAS #9012-36-6 (CHOCHO)n
Melting point: 87-89C

Agarose I is standard melting/gelling agarose, suitable for routine nucleic acid analytical/preservative applications. Agarose I has a low EEO for shorter electrophoresis runs without compromising resolution. Excellent for blotting technique and general manipulations.
pH: 5.7 7.7 (3%, Water) @ 25C

Catalog No. 50980167 Supplier Electron Microscopy Sciences Supplier No. 10205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS #9012-36-6 (CHOCHO)n
Melting point: 87-89C

Agarose I is standard melting/gelling agarose, suitable for routine nucleic acid analytical/preservative applications. Agarose I has a low EEO for shorter electrophoresis runs without compromising resolution. Excellent for blotting technique and general manipulations.
pH: 5.7 7.7 (3%, Water) @ 25C