Learn More
Thermo Scientific Chemicals Agarose MS8, molecular sieve grade, for small DNA fragments
For small DNA fragments | CAS: 9012-36-6
$342.47 - $990.34
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2673814
|
Thermo Scientific Chemicals
H2673814 |
25 g |
Each for $342.47
|
|
|||||
|
AAH2673822
|
Thermo Scientific Chemicals
H2673822 |
100 g |
Each for $990.34
|
|
|||||
Description
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAgarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 9012-36-6 | |
| MFCD00081294 |
Specifications
| 9012-36-6 | |
| Hygroscopic; Moisture sensitive | |
| Soluble in water. | |
| Agarose MS8, for small DNA fragments |
| 25 g | |
| MFCD00081294 | |
| Molecular Sieve |
Safety and Handling
EINECSNumber : 232-731-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only