missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Molecular Biology Grade, Hoefer™

Catalog No. GR140500X
Encompass
Change view
Click to view available options
Quantity:
25 g
500 g
2 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GR140500X 2 x 500 g
HBGR14025 25 g
03500523 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. GR140500X Supplier Hoefer Inc Supplier No. GR140 500X

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9012-36-6

Specifications

Physical Form Granular Free-flowing Powder
Quantity 2 x 500 g
Grade Molecular Biology
Chemical Name or Material Agarose