missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Medium-EEO/Protein Electrophoresis Grade), Fisher BioReagents

Catalog No. BP161100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP161100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP161100 Supplier Fisher BioReagents Supplier No. BP161 100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal alternative to polyacrylamide for serum protein electrophoresis. Immunoelectrophoresis. Electrophoresis of nucleic acids.

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 100 g
DNase DNase free
Electroendosmosis (EEO) 0.16 to 0.19
Gelation Temperature 34.5° to 37.5°C
Gel Strength 1000g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade Protein Electrophoresis
Chemical Name or Material Agarose, Medium-EEO
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT