Learn More
Thermo Scientific Chemicals Agarose ME, for electrophoresis of macromolecules
CAS: 9012-36-6
$83.49 - $274.68
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1669306
|
Thermo Scientific Chemicals
A1669306 |
5 g |
Each for $83.49
|
|
|||||
|
AAA1669314
|
Thermo Scientific Chemicals
A1669314 |
25 g |
Each for $274.68
|
|
|||||
Description
Agarose is frequently used in molecular biology for the separation of large molecules, especially DNA, by electrophoresis. Slabs of agarose gels (usually 0.7 - 2%) for electrophoresis are readily prepared by pouring the warm, liquid solution into a mold. A wide range of different agaroses, of varying molecular weights and properties are commercially available for this purpose. Used in high resolution protein electrophoresis, immunoelectrophoresis, electro immunoassays such as rocket assays, counter current electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAgarose is frequently used in molecular biology for the separation of large molecules, especially DNA, by electrophoresis. Slabs of agarose gels (usually 0.7 - 2%) for electrophoresis are readily prepared by pouring the warm, liquid solution into a mold. A wide range of different agaroses, of varying molecular weights and properties are commercially available for this purpose. Used in high resolution protein electrophoresis, immunoelectrophoresis, electro immunoassays such as rocket assays, counter current electrophoresis.
Solubility
Soluble in water (10 g/L at 80°C).
Notes
Gel point 36°. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Air, moisture.
Chemical Identifiers
| 9012-36-6 | |
| MFCD00081294 |
Specifications
| 9012-36-6 | |
| MFCD00081294 | |
| Agarose ME, for electrophoresis of macromolecules |
| 5 g | |
| Soluble in water (10g/L at 80°C). |
Safety and Handling
EINECSNumber : 232-731-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only