missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose ME, for electrophoresis of macromolecules

Catalog No. AAA1669314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1669314 25 g
AAA1669306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1669314 Supplier Thermo Scientific Chemicals Supplier No. A1669314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 9012-36-6

Agarose is frequently used in molecular biology for the separation of large molecules, especially DNA, by electrophoresis. Slabs of agarose gels (usually 0.7 - 2%) for electrophoresis are readily prepared by pouring the warm, liquid solution into a mold. A wide range of different agaroses, of varying molecular weights and properties are commercially available for this purpose. Used in high resolution protein electrophoresis, immunoelectrophoresis, electro immunoassays such as rocket assays, counter current electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is frequently used in molecular biology for the separation of large molecules, especially DNA, by electrophoresis. Slabs of agarose gels (usually 0.7 - 2%) for electrophoresis are readily prepared by pouring the warm, liquid solution into a mold. A wide range of different agaroses, of varying molecular weights and properties are commercially available for this purpose. Used in high resolution protein electrophoresis, immunoelectrophoresis, electro immunoassays such as rocket assays, counter current electrophoresis.

Solubility
Soluble in water (10 g/L at 80°C).

Notes
Gel point 36°. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Air, moisture.

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Quantity 25 g
Solubility Information Soluble in water (10g/L at 80°C).
Chemical Name or Material Agarose ME, for electrophoresis of macromolecules
EINECSNumber 232-731-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only