missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Low-Melting, <1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™

Catalog No. BP1360100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP1360100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP1360100 Supplier Fisher BioReagents Supplier No. FLBP1360100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agarose, C12H18O9, CAS Number-9012-36-6, 100g, Gray, Cream, DNase free, 0.15 max., Pass Test, 306.12, 35 deg.C max., 500g/cm2min., Genetic Analysis, Low-Melting, <1kb DNA/RNA, Poly Bottle, Powder/Solid, DNase- and RNase-Free, RT, 0.15% max. | CAS: 9012-36-6 | C12H18O9

Agarose, C12H18O9, CAS Number-9012-36-6, 100g, Gray, Cream, DNase free, 0.15 max., Pass Test, 306.12, 35 deg.C max., 500g/cm2min., Genetic Analysis, Low-Melting, <1kb DNA/RNA, Poly Bottle, Powder/Solid, DNase- and RNase-Free, RT, 0.15% max.

Specifications

Chemical Name or Material Agarose
Physical Form Powder/Solid
Name Note Low-Melting, <1kb DNA/RNA
CAS 9012-36-6
Color Cream
Quantity 100 g
DNase DNase free
Electroendosmosis (EEO) 0.15 max.
Gelation Temperature 35°C max.
Gel Strength 500g/cm2min.
Packaging Poly Bottle
Purity Grade Notes DNase- and RNase-Free
Molecular Formula C12H18O9
Formula Weight 306.12
Grade Genetic Analysis
Show More Show Less