missing translation for 'onlineSavingsMsg'
Learn More

Research Products Corporation Agarose, Low Melt Temperature, 50 Grams

Catalog No. 50213122
Encompass

Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.

Gel Strength, 1%: >200 g/cm2
Gelling Temperature: 26-30°C
Melting Temperature: <65°C
Sulfate: <0.10%
EEO: <0.10
DNase and RNase: None Detected

Catalog No. 50213122 Supplier Research Products Corporation Supplier No. RPI A2007050.0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.

Gel Strength, 1%: >200 g/cm2
Gelling Temperature: 26-30°C
Melting Temperature: <65°C
Sulfate: <0.10%
EEO: <0.10
DNase and RNase: None Detected