missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose low EEO, for electrophoresis, (Mr <= 0.12)

CAS: 9012-36-6

$472.82 - $472.82

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC159821000
SDS View Documents
Thermo Scientific Chemicals
159821000
100 g Glass Bottle
Each for $472.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Fine Crystalline Powder
White
electroendosmosis (-Mr): 0.07 to 0.12
>1800g/cm2 (1.5% gel)
Glass Bottle
Solubility in water: soluble in hot water. Other solubilities: insoluble in most common organic solvents
Agarose low EEO
9012-36-6
100 g
40 to 43°C (1.5% sol.)
Authentic
MFCD00081294
Electrophoresis
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only