missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, low EEO

Catalog No. p-7057484
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6636918 50 g
AAJ6636922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6636918 Supplier Thermo Scientific Chemicals Supplier No. J6636918
Only null left

CAS: 9012-36-6

Agarose is useful in separation of nucleic acids electrophoretically, Suitable for isoelectric focusing, protein blotting and antibody separation Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is useful in separation of nucleic acids electrophoretically, Suitable for isoelectric focusing, protein blotting and antibody separation Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

Solubility
Soluble in water

Notes
Gel strength: >2400g/cm² (1.5% w/v); Gelling temperature: 35-37°C; EEO: 0.07-0.1

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Melting Point 87°C to 89°C
Color White
Quantity 50 g
Sensitivity Hygroscopic; Moisture sensitive
Solubility Information Soluble in water
Chemical Name or Material Agarose, low EEO
EINECSNumber 232-731-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only