missing translation for 'onlineSavingsMsg'
Learn More

Agarose, LMP, Preparative Grade for Small Fragments (10 to 1000 bp), Promega

Catalog No. PRV3841
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3841 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV3841 Supplier Promega Corporation Supplier No. V3841

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Premium agarose used for isolating DNA fragments from 10 to 1,000bp.

  • Quality tested and certified free of DNase and RNase activity

Specifications

Chemical Name or Material Agarose LMP
Name Note For Small Fragments (10 to 1000 bp)
Physical Form Powder
Color White
Quantity 25 g
Grade Preparative