missing translation for 'onlineSavingsMsg'
Learn More

Agarose, LMP, Preparative Grade for Large Fragments (>1000 bp), Promega

Catalog No. PRV2831
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2831 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV2831 Supplier Promega Corporation Supplier No. V2831

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Premium agarose used for isolating DNA fragments larger than 1,000bp.

  • Quality tested and certified free of DNase and RNase activity

Specifications

Chemical Name or Material Agarose LMP
Name Note For Large Fragments (>1000 bp)
Physical Form Powder
Color White
Quantity 25 g
Grade Preparative