missing translation for 'onlineSavingsMsg'
Learn More

Analytical Grade Agarose LE, Promega

Catalog No. PRV3121
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3121 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV3121 Supplier Promega Corporation Supplier No. V3121

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for electrophoretic separation of nucleic acids.

  • Quality tested and certified free of DNase and RNase activity

Specifications

Chemical Name or Material Agarose LE
Physical Form Powder
Name Note For the electrophoretic separation of nucleic acids
Color White
Quantity 100 g
Grade Analytical