missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Intermediate Melting/PCR Grade), Fisher BioReagents

Catalog No. BP2410100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP2410100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP2410100 Supplier Fisher BioReagents Supplier No. BP2410100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For separation of low MW DNA ≤1000 bp by PCR analysis

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 100 g
DNase DNase free
Electroendosmosis (EEO) 0.12 max.
Gelation Temperature 35.5°C max.
Gel Strength 1200g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade DNase- and RNase-Free
Chemical Name or Material Agarose, Intermediate Melting
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT