missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Genetic Technology Grade, MP Biomedicals™

$1782.88 - $1782.88

Chemical Identifiers

CAS 9012-36-6
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN800669
View Documents
MP Biomedicals Inc
04800669
500 g
Each for $1,782.88
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9012-36-6

Specifications

9012-36-6
500 g
Genetic Technology
Agarose
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'