missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, Genetic Technology Grade

CAS: 9012-36-6

$268.33 - $577.26

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6620814
View Documents
Thermo Scientific Chemicals
J6620814
25 g
Each for $268.33
Only null left
 
AAJ6620822
View Documents
Thermo Scientific Chemicals
J6620822
100 g
Each for $577.26
Only null left
 
Description

Description

Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes
Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Powder
White
Hygroscopic; Moisture sensitive
Genetic Technology
9012-36-6
25 g
MFCD00081294
Agarose
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only