missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)

$133.11 - $133.11

Specifications

Chemical Name or Material Agarose gel loading dye
Physical Form Liquid
Name Note glycerol based, Xylene Cyanol FF free
Concentration 6X
Solubility Information Fully misicible in water.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60933AC
View Documents
Thermo Scientific Chemicals
J60933AC
25 mL
Each for $133.11
Only null left
 
Description

Description

6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

Solubility
Fully misicible in water.

Notes
No Xylene cyanol FF. Contains 0.25% bromophenol blue, 30% glycerol.
Specifications

Specifications

Agarose gel loading dye
glycerol based, Xylene Cyanol FF free
Fully misicible in water.
Liquid
6X
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only